PRIH30452866 Princeton BioMolecular Research, Inc.

3-Amino-4,6-dimethyl-furo[2,3-b]pyridine-2-carboxylic acid methyl ester

  • MDL number MFCD06011068
  • Cas Number : 801228-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart