PRIH3045286D Princeton BioMolecular Research, Inc.

3-(methylthio)-4-oxo-4,5,6,7-tetrahydro-2-benzothiophene-1-carbonitrile

  • MDL number MFCD00084985
  • Cas Number : 168279-53-6
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart