PRIH30A13428 Princeton BioMolecular Research, Inc.

4-(3-Piperidin-3-yl-[1,2,4]oxadiazol-5-yl)-pyridine

  • MDL number MFCD18054530
  • Cas Number : 915277-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart