PRIH30A1343A Princeton BioMolecular Research, Inc.

4-cyclopropyl-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD22030969
  • Cas Number : 1369139-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart