PRIH30A1343D Princeton BioMolecular Research, Inc.

dimethyl 6-chloropyridine-3,4-dicarboxylate

  • MDL number MFCD28972751
  • Cas Number : 1189570-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart