PRIH30A1343E Princeton BioMolecular Research, Inc.

2-cyclopropyl-2-oxoethyl acetate

  • MDL number MFCD24393396
  • Cas Number : 89898-26-0
  • Molecular Weight : 142.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart