PRIH30A13444 Princeton BioMolecular Research, Inc.

1-(2,2,2-trifluoroethyl)indol-4-amine

  • MDL number MFCD20282162
  • Cas Number : 1353949-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart