PRIH30A13445 Princeton BioMolecular Research, Inc.

2,6-bis(trifluoromethyl)pyrimidin-4-ol

  • MDL number MFCD08436595
  • Cas Number : 884-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart