PRIH3239D4C7 Princeton BioMolecular Research, Inc.

1-isobutyl-1H-indol-5-amine

  • MDL number MFCD11213924
  • Cas Number : 1094710-32-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart