PRIH3239D52D Princeton BioMolecular Research, Inc.

(6-bromo-1H-indol-3-yl)acetic acid

  • MDL number MFCD09954810
  • Cas Number : 152213-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart