PRIH440C8B4B Princeton BioMolecular Research, Inc.

3,4-Dichloro-benzo[d]isoxazole

  • MDL number MFCD18378856
  • Cas Number : 1344687-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart