PRIH440C8B4C Princeton BioMolecular Research, Inc.

4-Phenyl-cyclohexanol

  • MDL number MFCD00013757
  • Cas Number : 5437-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart