PRIH440C8B4D Princeton BioMolecular Research, Inc.

3-(2-Methoxy-ethyl)-2,4-dioxo-1,3-diaza-spiro[4.5]decane-8-carboxylic acid

  • MDL number MFCD26381954
  • Cas Number : 1708427-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart