PRIH440C8B5B Princeton BioMolecular Research, Inc.

2-(4-cyclopropyl-1H-1,2,3-triazol-1-yl)ethanamine

  • MDL number MFCD26881585
  • Cas Number : 1566055-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart