PRIH440C8B5F Princeton BioMolecular Research, Inc.

2-(5-bromopyridin-3-yl)-2,2-dimethoxyethanamine

  • MDL number MFCD18259240
  • Cas Number : 1256785-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart