PRIH440C8B64 Princeton BioMolecular Research, Inc.

methyl [4-(1,1-dimethylpropyl)-1H-1,2,3-triazol-1-yl]acetate

  • MDL number MFCD31919106
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart