PRIH440C8B66 Princeton BioMolecular Research, Inc.

1-(2-bromophenyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD10686844
  • Cas Number : 1039325-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart