PRIH440C8B67 Princeton BioMolecular Research, Inc.

2-methyl-2-(1-methyl-1H-imidazol-2-yl)propanoic acid

  • MDL number MFCD19621770
  • Cas Number : 865157-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart