PRIH440C8B69 Princeton BioMolecular Research, Inc.

2-{1-[(1R,2R)-2-aminocyclohexyl]-1H-1,2,3-triazol-4-yl}propan-2-ol

  • MDL number MFCD31919119
  • Molecular Weight : 224.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart