PRIH8BA055AE Princeton BioMolecular Research, Inc.

1-(5-chloro-2-nitrophenyl)-1H-imidazole

  • MDL number MFCD06795458
  • Cas Number : 190198-20-0
  • Molecular Weight : 223.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart