PRIH8BA055DB Princeton BioMolecular Research, Inc.

[3-(trifluoromethyl)-1H-pyrazol-1-yl]acetyl chloride

  • MDL number MFCD11106712
  • Cas Number : 1174311-36-4
  • Molecular Weight : 212.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart