PRIH8BA0560C Princeton BioMolecular Research, Inc.

(2-Amino-4-hydroxy-6-methyl-pyrimidin-5-yl)-acetic acid

  • MDL number MFCD03815450
  • Cas Number : 166267-96-5
  • Molecular Weight : 183.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart