PRIH8BA05658 Princeton BioMolecular Research, Inc.

5-hydroxy-5-methyl-3-(4-methylphenyl)cyclohex-2-en-1-one

  • MDL number MFCD12403060
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart