PRIH8BA0565C Princeton BioMolecular Research, Inc.

(3-fluoro-4-methoxyphenyl)methanesulfonyl chloride

  • MDL number MFCD11226947
  • Molecular Weight : 238.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart