PRIH8BA056C9 Princeton BioMolecular Research, Inc.

2-Methyl-2-pyridin-3-yl-propionic acid

  • MDL number MFCD13563031
  • Cas Number : 169253-35-4
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart