PRIH8BA056E0 Princeton BioMolecular Research, Inc.

methyl 1-(4-chlorophenyl)-3-oxocyclobutanecarboxylate

  • MDL number MFCD22543820
  • Cas Number : 1370618-03-3
  • Molecular Weight : 238.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart