PRIH8BA056E8 Princeton BioMolecular Research, Inc.

4-amino-N-butyl-1-ethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD04969500
  • Cas Number : 1006484-73-6
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart