PRIH8BA056EA Princeton BioMolecular Research, Inc.

3-amino-N-ethyl-1-methyl-N-phenyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD15976701
  • Cas Number : 2101197-52-6
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart