PRIH8BA056EF Princeton BioMolecular Research, Inc.

methyl 1-(2,4-dichlorobenzyl)-3-(difluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD31417554
  • Cas Number : 2054954-63-9
  • Molecular Weight : 335.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart