PRIH8BA056FF Princeton BioMolecular Research, Inc.

4-bromo-1-isobutyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD31417579
  • Cas Number : 1784836-38-9
  • Molecular Weight : 231.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart