PRIH8BA05704 Princeton BioMolecular Research, Inc.

4-iodo-1-phenyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD31417584
  • Cas Number : 2054953-44-3
  • Molecular Weight : 298.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart