PRIH8BA05707 Princeton BioMolecular Research, Inc.

3-(4-nitro-1'H-1,4'-bipyrazol-1'-yl)butanoic acid

  • MDL number MFCD31417588
  • Cas Number : 2054953-25-0
  • Molecular Weight : 265.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart