PRIH8BA05761 Princeton BioMolecular Research, Inc.

ethyl 3-[3-(difluoromethyl)-1H-pyrazol-1-yl]-4,4,4-trifluorobutanoate

  • MDL number MFCD31417617
  • Cas Number : 2054953-10-3
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart