PRIH8BA05766 Princeton BioMolecular Research, Inc.

ethyl 4,4,4-trifluoro-3-(3-methoxy-4-nitro-1H-pyrazol-1-yl)butanoate

  • MDL number MFCD31417622
  • Cas Number : 2054953-09-0
  • Molecular Weight : 311.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart