PRIH8BA0576A Princeton BioMolecular Research, Inc.

4,4,4-trifluoro-3-(3-methyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD20329368
  • Cas Number : 1342567-97-8
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart