PRIH8BA05775 Princeton BioMolecular Research, Inc.

3-[3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]-4,4,4-trifluorobutanoic acid

  • MDL number MFCD31417634
  • Cas Number : 2054953-73-8
  • Molecular Weight : 272.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart