PRIH8BA0577A Princeton BioMolecular Research, Inc.

4,4,4-trifluoro-3-(3-phenyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD31417637
  • Cas Number : 2054953-40-9
  • Molecular Weight : 284.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart