PRIH8BA05781 Princeton BioMolecular Research, Inc.

3-amino-1-ethyl-N-isobutyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31417730
  • Cas Number : 2101199-95-3
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart