PRIH8BA05796 Princeton BioMolecular Research, Inc.

3-amino-N-(2-furylmethyl)-1-isopropyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31417751
  • Cas Number : 2101198-27-8
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart