PRIH8BA057AA Princeton BioMolecular Research, Inc.

1-isopropyl-5-(piperidin-1-ylcarbonyl)-1H-pyrazol-3-amine

  • MDL number MFCD31417763
  • Cas Number : 1891178-52-1
  • Molecular Weight : 236.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart