PRIH8BA057B2 Princeton BioMolecular Research, Inc.

3-amino-N-ethyl-N,1-dimethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD31417771
  • Cas Number : 2101198-37-0
  • Molecular Weight : 182.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart