PRIH8BA057B6 Princeton BioMolecular Research, Inc.

3-amino-1-methyl-N-(3-morpholin-4-ylpropyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD31417775
  • Cas Number : 2101200-77-3
  • Molecular Weight : 267.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart