PRIH8BA057BB Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-N,1-dimethyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD21194585
  • Cas Number : 1468545-93-8
  • Molecular Weight : 182.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart