PRIH8BA057BE Princeton BioMolecular Research, Inc.

4-amino-N-[2-(dimethylamino)ethyl]-1-methyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD21221394
  • Cas Number : 1492976-49-4
  • Molecular Weight : 211.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart