PRIH8BA057CF Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-1-methyl-N-phenyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD20998361
  • Cas Number : 1487783-80-1
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart