PRIH8BA057E9 Princeton BioMolecular Research, Inc.

4-amino-N-butyl-1-isopropyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417786
  • Cas Number : 2101198-58-5
  • Molecular Weight : 224.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart