PRIH8BA05808 Princeton BioMolecular Research, Inc.

4-amino-N-cyclohexyl-1-isopropyl-N-methyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417811
  • Cas Number : 2101199-53-3
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart