PRIH8BA05812 Princeton BioMolecular Research, Inc.

4-amino-N-ethyl-N-methyl-1-propyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417821
  • Cas Number : 2101197-65-1
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart