PRIH8BA05816 Princeton BioMolecular Research, Inc.

4-amino-N-cyclohexyl-1-propyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD31417825
  • Cas Number : 2101198-31-4
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart