PRIH8BA05826 Princeton BioMolecular Research, Inc.

3-[(3-methylpiperidin-1-yl)carbonyl]-1-propyl-1H-pyrazol-4-amine

  • MDL number MFCD31417841
  • Cas Number : 2101195-67-7
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart